Anti-AP1G1 Rabbit Polyclonal Antibody
Catalog # USBIA2298-48B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:adaptor-related protein complex 1, gamma 1 subunit
- Antigen symbol:AP1G1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Format:Serum
- Form:lyophilized powder
- Amino acid number:5 to 24
- Storage buffer:100ul sterile ddH2O.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000-1:4000; Immunochemistry: 1:200-1:2000
Type: Primary
Antigen: AP1G1 (adaptor-related protein complex 1, gamma 1 subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat