Anti-AK1 Rabbit Polyclonal Antibody (Biotin)
Catalog # USBIA0882-65B-BIOT
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:adenylate kinase 1
- Antigen symbol:AK1
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Hamster,Human
- Western blot:Yes
- Epitope:C-terminal
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: AK1 (adenylate kinase 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Hamster, Human