Order Entry
Poland
ContactUsLinkComponent
Anti-ADAMTS17 Mouse Monoclonal Antibody [clone: 3B7]
Anti-ADAMTS17 Mouse Monoclonal Antibody [clone: 3B7]
Catalog # USBIA0859-67A
Supplier:  US Biological
undefined
Anti-ADAMTS17 Mouse Monoclonal Antibody [clone: 3B7]
Catalog # USBIA0859-67A
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ADAM metallopeptidase with thrombospondin type 1 motif, 17
  • Antigen symbol:
    ADAMTS17
  • Clonality:
    Monoclonal
  • Clone:
    3B7
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Amino acid number:
    543 to 651
  • Storage buffer:
    PBS, pH 7,2.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Recommended dilution: Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: ADAMTS17 (ADAM metallopeptidase with thrombospondin type 1 motif, 17)
Clonality: monoclonal
Clone: 3B7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human