Order Entry
Poland
ContactUsLinkComponent
Anti-ADAM 32 Sheep Polyclonal Antibody
Anti-ADAM 32 Sheep Polyclonal Antibody
Catalog # USBIA0859-41M
Supplier:  US Biological
Anti-ADAM 32 Sheep Polyclonal Antibody
Catalog # USBIA0859-41M
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ADAM metallopeptidase domain 32
  • Antigen symbol:
    ADAM 32
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Sheep
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    lyophilized powder
  • Amino acid number:
    17 to 476
  • Storage buffer:
    0,2um sterile-filtered solution, PBS, 5% trehalose.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,2 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: ELISA: 0.5-1.0 ?g/ml; Western Blot: 1.0 ?g/ml

Type: Primary
Antigen: ADAM 32
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human