Order Entry
Poland
ContactUsLinkComponent
Anti-14-3-3 gamma Mouse Monoclonal Antibody [clone: 9A70]
Anti-14-3-3 gamma Mouse Monoclonal Antibody [clone: 9A70]
Catalog # USBI0014-07E
Supplier:  US Biological
Anti-14-3-3 gamma Mouse Monoclonal Antibody [clone: 9A70]
Catalog # USBI0014-07E
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    14-3-3 gamma
  • Clonality:
    Monoclonal
  • Clone:
    9A70
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Format:
    ascites
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: 14-3-3 gamma (14-3-3 gamma)
Clonality: Monoclonal
Clone: 9A70
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human