Order Entry
Poland
ContactUsLinkComponent
 
Anti-14-3-3 epsilon Mouse Monoclonal Antibody [clone: 5A5]
  USBI0014-04G
Anti-14-3-3 epsilon Mouse Monoclonal Antibody [clone: 5A5]
  USBI0014-04G

 

  • Antibody type:
    Primary
  • Antigen symbol:
    14-3-3 epsilon
  • Clonality:
    Monoclonal
  • Clone:
    5A5
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Amino acid number:
    1 to 255
  • Storage buffer:
    PBS, pH 7,4, 0,1% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

 

 

Recommended dilution: Western Blot: 1:1000-1:3000; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: 14-3-3 epsilon (14-3-3 epsilon)
Clonality: Monoclonal
Clone: 5A5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human