Anti-Troponin T Mouse Monoclonal Antibody [clone: TT-98]
Catalog # USBI145325
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Troponin T
- Clonality:Monoclonal
- Clone:TT-98
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Rabbit,Chicken,Rat
- Western blot:Yes
- Format:ascites
- Form:lyophilized powder
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Troponin T
Clonality: monoclonal
Clone: TT-98
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Rabbit, Rat