Anti-S-100, beta Mouse Monoclonal Antibody [clone: SA-12]
USBI145139
- Antibody type:Primary
- Antigen symbol:S-100, beta
- Clonality:Monoclonal
- Clone:SA-12
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Rabbit,Human,Rat
- Western blot:Yes
- Format:ascites
- Form:lyophilized powder
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: S-100, beta
Clonality: monoclonal
Clone: SA-12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rabbit, Rat