Order Entry
Poland
Orders LinkContactUsLinkComponent
Anti-BAK1 Rabbit Polyclonal Antibody | Avantor

Anti-BAK1 Rabbit Polyclonal Antibody

100 µg
numer katalogowy USBI144221
Dostawca:   US Biological
Anti-BAK1 Rabbit Polyclonal Antibody
numer katalogowy USBI144221
Dostawca:  US Biological
Anti-BAK1 Rabbit Polyclonal Antibody

100 µg

Przejdź do:
Price
2 150 zł
Zaloguj się dla dostępności
1 * 100 µG
Product Details & Documents

Type: Primary
Antigen: BAK1 (BCL2-antagonist/killer 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Documents

Dane techniczne
  • Size:
    100 µg
  • Western blot:
    Yes
  • Clonality:
    Polyclonal
  • Purification:
    purified by immunoaffinity chromatography
  • Antibody type:
    Primary
  • Form:
    lyophilized powder
  • Isotype:
    IgG
  • Storage temperature:
    Cool
  • Host:
    Rabbit
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Antigen name:
    BCL2-antagonist/killer 1
  • Cat. No.:
    USBI144221
  • Reactivity:
    Human|Rat|Mouse
  • ImmunoChemistry:
    Yes
  • Conjugation:
    Unconjugated
  • Antigen symbol:
    BAK1
  • Storage buffer:
    5mgBSA, 0,9mg NaCl, 0,2mg Na2HPO4, 0,05mgthimerosal, 0,05mg NaN3.