Order Entry
Poland
ContactUsLinkComponent
Anti-UDP Glucose Ceramide Glucosyltransferase Rabbit Polyclonal Antibody
Anti-UDP Glucose Ceramide Glucosyltransferase Rabbit Polyclonal Antibody
  USBI142720-50
Anti-UDP Glucose Ceramide Glucosyltransferase Rabbit Polyclonal Antibody
  USBI142720-50

 

  • Antibody type:
    Primary
  • Antigen name:
    UDP Glucose Ceramide Glucosyltransferase
  • Antigen symbol:
    UGCG
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2, 0,05% sodium azide, 50% glycerol.
  • Storage temperature:
    Cool
  • Concentration:
    200 ug/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

 

 

Recommended dilution: ELISA: 1:100-200; Western Blot: 1:100-400; Immunochemistry: 1:100-500

Type: Primary
Antigen: UDP Glucose Ceramide Glucosyltransferase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human