Anti-Semaphorin 3A Rabbit Polyclonal Antibody
USBI142243-50
- Antibody type:Primary
- Antigen name:Semaphorin 3A
- Antigen symbol:SEMA3A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,05% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Concentration:200 ug/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Recommended dilution: ELISA: 1:100-200; Western Blot: 1:100-400; Immunochemistry: 1:100-500
Type: Primary
Antigen: Semaphorin 3A
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human