Anti-Major Histocompatibility Complex Class II DR Alpha Mouse Monoclonal Antibody [clone: 13j65]
Catalog # USBI141369
Dostawca: US Biological
Dane techniczne
- Antibody type:Primary
- Antigen name:Major Histocompatibility Complex Class 2 DR Alpha
- Clonality:Monoclonal
- Clone:13j65
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Monkey
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,05% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:200 µg
- Pk:200 µG
Dane techniczne
Informacje o tym produkcie
Recommended dilution: ELISA: 1:100-200; Western Blot: 1:100-400; Immunochemistry: 1:100-500
Type: Primary
Antigen: Major Histocompatibility Complex Class II DR Alpha
Clonality: monoclonal
Clone: 13j65
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Monkey