Order Entry
Poland
ContactUsLinkComponent
Anti-NR2B Mouse Monoclonal Antibody [clone: S59-20]
Anti-NR2B Mouse Monoclonal Antibody [clone: S59-20]
Catalog # USBI136831
Supplier:  US Biological
Anti-NR2B Mouse Monoclonal Antibody [clone: S59-20]
Catalog # USBI136831
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    NR2B
  • Clonality:
    Monoclonal
  • Clone:
    S59-20
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Amino acid number:
    20 to 271
  • Storage buffer:
    PBS, pH 7,4, 50% glycerol.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: Immunofluorescence: 1-10 ?g/ml; Western Blot: 1 ?g/ml; Immunochemistry: 0.1-1 ?g/ml;

Type: Primary
Antigen: NR2B
Clonality: monoclonal
Clone: S59-20
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat