Anti-Synaptotagmin 11 Mouse Monoclonal Antibody [clone: 4E1]
Catalog # USBI134132
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Synaptotagmin 11
- Clonality:Monoclonal
- Clone:4E1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:84 to 140
- Storage buffer:PBS, pH 7,2.
- Sequence:GPGREGGRRNLLVDAAEAGLLSRDKDPRGPSSGSCIDQLPIKMDYGEELRSPITSL*
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunofluorescence: 10 ?g/ml
Type: Primary
Antigen: Synaptotagmin 11
Clonality: monoclonal
Clone: 40
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human