Anti-S100A13 Mouse Monoclonal Antibody [clone: 3A7]
USBI132918
- Antibody type:Primary
- Antigen name:S100 Calcium Binding Protein A13
- Antigen symbol:S100A13
- Clonality:Monoclonal
- Clone:3A7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:1 to 99
- Storage buffer:PBS, pH 7,2.
- Sequence:MAAEPLTELEESIETVVTTFFTFARQEGRKDSLSVNEFKELVTQQLPHLLKDVGSLDEKMKSLDVNQDSELKFNEYWRLIGELAKEIRKKKDLKIRKK*
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Immunofluorescence: 10 ?g/ml
Type: Primary
Antigen: S100A13 (S100 calcium binding protein A13)
Clonality: monoclonal
Clone: 3A7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human