Anti-RBAK Mouse Monoclonal Antibody [clone: 6F9]
USBI132367
- Antibody type:Primary
- Antigen name:RB-associated KRAB zinc finger
- Antigen symbol:RBAK
- Clonality:Monoclonal
- Clone:6F9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Amino acid number:51 to 151
- Storage buffer:PBS, pH 7,2.
- Sequence:TTKPNVIIKLEQGEEPWIMGGEFPCQHSPEAWRVDDLIERIQENEDKHSRQAACINSKTLTEEKENTFSQIYMSLVPSSIIAHNCVSCGKNLESISQL
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: RBAK (RB-associated KRAB zinc finger)
Clonality: monoclonal
Clone: 6F9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human