Anti-LUM Rabbit Polyclonal Antibody
Catalog # USBI129248
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Lumican
- Antigen symbol:LUM
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:1 to 338
- Storage buffer:PBS, pH 7,2.
- Sequence:MSLSAFTLFLALIGGTSGQYYDYDFPLSIYGQSSPNCAPECNCPESYPSAMYCDELKLKSVPMVPPGIKYLYLRNNQIDHIDEKAFENVTDLQWLILDHNLLENSKIKGRVFSKLKQLKKLHINHNNLTESVGPLPKSLEDLQLTHNKITKLGSFEGLVNLTFIHLQHNRLKEDAVSAAFKGLKSLEYLDLSFNQIARLPSGLPVSLLTLYLDNNKISNIPDEYFKRFNALQYLRLSHNELADSGIPGNSFNVSSLVELDLSYNKLKNIPTVNENLENYYLEVNQLEKFDIKSFCKILGPLSYSKIKHLRLDGNRISETSLPPDMYECLRVANEVTLN
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: LUM (lumican)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human