Anti-HDAC3 Mouse Monoclonal Antibody [clone: 2A3]
Catalog # USBI127763
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Histone deacetylase 3
- Antigen symbol:HDAC3
- Clonality:Monoclonal
- Clone:2A3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Amino acid number:319 to 429
- Storage buffer:PBS, pH 7,2.
- Sequence:ISEELPYSEYFEYFAPDFTLHPDVSTRIENQNSRQYLDQIRQTIFENLKMLNHAPSVQIHDVPADLLTYDRTDEADAEERGPEENYSRPEAPNEFYDGDHDNDKESDVEI
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunofluorescence: 10 ?g/ml
Type: Primary
Antigen: HDAC3 (histone deacetylase 3)
Clonality: monoclonal
Clone: 2A3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse, Rat