Anti-CSTF1 Mouse Monoclonal Antibody [clone: 1C6]
Catalog # USBI125423
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:cleavage stimulation factor, 3' pre-RNA, subunit 1, 50kDa
- Antigen symbol:CSTF1
- Clonality:Monoclonal
- Clone:1C6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:332 to 431
- Storage buffer:PBS, pH 7,2.
- Sequence:LWEISTGRTLVRYTGAGLSGRQVHRTQAVFNHTEDYVLLPDERTISLCCWDSRTAERRNLLSLGHNNIVRCIVHSPTNPGFMTCSDDFRARFWYRRSTTD
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: CSTF1 (cleavage stimulation factor, 3' pre-RNA, subunit 1, 50kDa)
Clonality: monoclonal
Clone: 1C6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human