Anti-ABCC11 Rabbit Polyclonal Antibody
Catalog # USBI122803
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:ATP Binding Cassette, sub-family C, member 11
- Antigen symbol:ABCC11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Format:Serum
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: ABCC11 (ATP-binding cassette, sub-family C (CFTR/MRP), member 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human