Anti-Caspase-4/11, p20 Mouse Monoclonal Antibody [clone: Flamy-1]
Catalog # USBI044637
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Caspase 4/11, p20
- Antigen symbol:CASP4
- Clonality:Monoclonal
- Clone:Flamy-1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2b kappa
- Western blot:Yes
- Form:liquid
- Amino acid number:81 to 373
- Storage buffer:PBS, 10% glycerol, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Caspase-4/11, p20
Clonality: Monoclonal
Clone: Flamy-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: