Anti-MIIP Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Catalog # USBI038436-AP
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:migration and invasion inhibitory Protein
- Antigen symbol:MIIP
- Clonality:Polyclonal
- Conjugation:AP (Alkaline Phosphatase)
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Form:liquid
- Amino acid number:280 to 320
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000
Do not freeze!
Type: Primary
Antigen: MIIP (migration and invasion inhibitory protein)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human