Anti-DMRTC2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # USBI034686-FITC
Supplier: US Biological
Specifications
- Typ przeciwciała:Primary
- Nazwa antygenu:DMRT-like family C2
- Symbol antygenu:DMRTC2
- Klonalność:Poliklonalne
- Koniugacja:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Gospodarz:Rabbit
- Izotyp:IgG
- Reaktywność:Human
- Western blot:Yes
- Epitop:internal
- Formularz:liquid
- Liczba aminokwasów:98 to 128
- Bufor do przechowywania:PBS, pH 7,2, contains ~0,09% sodium azide
- Temperatura przechowywania:Cool
- Temperatura transportu:Ice
- Oczyszczenie:purified by Protein A chromatography
- Rozmiar:200 µl
- Ilość:200 µl
Specifications
About this item
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: DMRTC2 (DMRT-like family C2)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human