Order Entry
Poland
ContactUsLinkComponent
Anti-CD3 Mouse Monoclonal Antibody [clone: OKT3]
Anti-CD3 Mouse Monoclonal Antibody [clone: OKT3]
  USBI030846
undefined
Anti-CD3 Mouse Monoclonal Antibody [clone: OKT3]
  USBI030846

 

  • Antibody type:
    Primary
  • Antigen symbol:
    CD3
  • Clonality:
    Monoclonal
  • Clone:
    OKT3
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2, 0,09% sodium azide
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    200 µl
  • Pk:
    200 µl

 

 

Recommended dilution: Flow Cytometry: 1:100

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: OKT3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human