Order Entry
Poland
ContactUsLinkComponent
Anti-AP2M1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-AP2M1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # USBI029029
Supplier:  US Biological
undefined
Anti-AP2M1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # USBI029029
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    adaptor-related protein complex 2, mu 1 subunit
  • Antigen symbol:
    AP2M1
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    0,01M PBS, PH 7.4, 0,03% Procl300, 50% glycerol
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by ammonium sulfate precipitation
  • Size:
    50 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: AP2M1 (adaptor-related protein complex 2, mu 1 subunit)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human