Anti-IgG Goat Polyclonal Antibody
ROCK618-101-012
- Antibody type:Secondary
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Ferret
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Storage buffer:0,125 M Sodium Borate, 0,075 M Sodium Chloride, 0,005 M EDTA, pH 8.0, 0,01% (w/v) Sodium Azide
- Storage temperature:Cool
- Concentration:1,0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Size:1 mg
- Pk:1 mg
Type: Secondary
Antigen: IgG γ
Clonality:
Clone:
Conjugation: Unconjugated
Epitope: γ chain
Host:
Isotype:
Reactivity: Ferret