Anti-IgG gamma Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Catalog # ROCK618-100-012
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG gamma
- Clonality:Polyclonal
- Conjugation:TRITC (Tetramethylrhodamine Isothiocyanate)
- Host:Goat
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Ferret
- Environmentally Preferable:
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope:
Host: Goat
Isotype:
Reactivity: Ferret