Order Entry
Poland
ContactUsLinkComponent
Anti-IgG1 Rabbit Polyclonal Antibody (Peroxidase)
Anti-IgG1 Rabbit Polyclonal Antibody (Peroxidase)
Catalog # ROCK610-4340
undefined
Anti-IgG1 Rabbit Polyclonal Antibody (Peroxidase)
Catalog # ROCK610-4340

Specifications

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG1
  • Clonality:
    Polyclonal
  • Conjugation:
    Peroxidase
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Gel shift:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • ChIP:
    Yes
  • Neutralisation:
    Yes
  • Format:
    Antibody fraction
  • Form:
    Lyophilized
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Gentamicin Sulfate. 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
  • Storage temperature:
    Cool
  • Concentration:
    1,0 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Ice
  • Immunogen:
    Mouse IgG1 heavy chain
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    1 mg
  • Pk:
    1 mg

Specifications

About this item

Secondary Rabbit Anti- IgG1 Reacts with Mouse

Recommended Dilutions: ELISA: 1:20000 - 1:200,000; Western Blot: 1:2000 - 1:20000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!

Type: Secondary
Antigen: IgG1 γ
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse