Order Entry
Poland
ContactUsLinkComponent
Anti-IgG F(ab')2 Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-IgG F(ab')2 Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  ROCK610-1204
Anti-IgG F(ab')2 Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  ROCK610-1204

 

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG F(ab')2
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Gel shift:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • ChIP:
    Yes
  • Neutralisation:
    Yes
  • Format:
    Antibody fraction
  • Form:
    Lyophilized
  • Storage buffer:
    0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
  • Storage temperature:
    Cool
  • Concentration:
    2,0 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Ice
  • Immunogen:
    Mouse IgG F(ab')2 fragment
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    2 mg
  • Pk:
    2 mg

 

 

Secondary Goat Anti-IgG F(ab')2 Reacts with Mouse

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope: F(ab')2
Host: Goat
Isotype: IgG
Reactivity: Mouse