Order Entry
Poland
ContactUsLinkComponent
Anti-IgG (H+L) Rabbit Polyclonal Antibody
Anti-IgG (H+L) Rabbit Polyclonal Antibody
Catalog # ROCK216-4102
Anti-IgG (H+L) Rabbit Polyclonal Antibody
Catalog # ROCK216-4102

Specifications

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG (H+L)
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Gel shift:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
  • Reactivity:
    Donkey
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    none
  • ChIP:
    Yes
  • Neutralisation:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 mg
  • Pk:
    50 mg

Specifications

About this item

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Donkey