Anti-IgM µ Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
numer katalogowy ROCK213-4307
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Secondary
- Antigen symbol:IgM µ
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Flow cytometry:Yes
- Gel shift:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Sheep
- Western blot:Yes
- Cross adsorption:none
- ChIP:Yes
- Neutralisation:Yes
- Format:Antibody fraction
- Form:Lyophilized
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Gentamicin Sulfate. 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
- Storage temperature:Cool
- Concentration:10,0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:Sheep IgM mu heavy chain
- Purification:multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by cross adsorption and extensive dialysis against the buffer stated above
- Size:20 mg
- Pk:20 mg
Specifications
About this item
This product has been assayed against 1.0 µg of Sheep IgM in a standard capture ELISA using ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid)) as a substrate for 30 minutes at room temperature.
Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:1000 - 1:10000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 2.0 ml
Do not add Sodium Azide!
Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: µ-chain
Host: Rabbit
Isotype: IgM
Reactivity: Sheep