Anti-Donkey Serum Rabbit Polyclonal Antibody
ROCK116-4101
- Antibody type:Primary
- Antigen name:Donkey Serum
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Donkey
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Format:Antiserum
- Form:Lyophilized
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:90 mg/ml by Refractometry
- Shipping temperature:Ice
- Immunogen:Donkey serum Proteins
- Purification:delipidation and defibrination
- Size:2 ml
- Pk:2 mL
Primary Rabbit Anti-DONKEY SERUM Reacts with Donkey
Recommended Dilutions:
Restore with deionized water (or equivalent); 2.0 ml
Type: Primary
Antigen: Donkey Serum
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Donkey