Anti-PIK3CD Rabbit Polyclonal Antibody
numer katalogowy ROCK100-401-862
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:phosphatidylinositol 3-kinase catalytic delta polypeptide
- Antigen symbol:PIK3CD
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Format:Antiserum
- Form:Liquid (sterile filtered)
- Gene ID:18707
- Antigen synonyms:2610208K16Rik|p110delta|AW545373|2410099E07Rik
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide
- Storage temperature:Cool
- Concentration:85 mg/ml by Refractometry
- Shipping temperature:Ice
- Immunogen:This antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a region near the C-terminal of mouse PI3K p110d. This sequence is identical in both mouse and human.
- Purification:delipidation and defibrination
- Size:100 µl
- Pk:100 µl
Specifications
About this item
This antibody has been tested for use in ELISA, western blotting and immunoprecipitation. Reactivity in other immunoassays is unknown. A mouse whole cell splenic lysate is suitable for use as a positive control.
Recommended Dilutions: ELISA: 1:4000 - 1:20000; Western Blot: 1:5000;
Type: Primary
Antigen: PIK3CD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Mouse, Human