Anti-CREB1 Rabbit Polyclonal Antibody
100 µl
Catalog # ROCK100-401-195
Supplier: Rockland Immunochemicals
Jump to:
Product Details & Documents
Anti-CREB-1 Antibody is suitable for use in SDS, Western Blot, and ELISA assays. Specific conditions for reactivity should be optimized by the end user.
Recommended Dilutions: ELISA: 1:5000 - 1:25000; Gel Shift: 0.5 to 1.0 µl per assay; Western Blot: 1:500 - 1:1000;
Type: Primary
Antigen: CREB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat, Human
Documents
Specifications
- Size:100 µl
- Western blot:Yes
- Clonality:Polyclonal
- ImmunoPrecipitation:Yes
- Purification:delipidation and immunoadsorption against an E.coli lysate immobilized on agarose beads
- Antibody type:Primary
- Epitope:near the N-terminus
- Form:Liquid (sterile filtered)
- Storage temperature:Cool
- Host:Rabbit
- Concentration:2,56 mg/ml by UV absorbance at 280 nm
- Cross adsorption:none
- Shipping temperature:Ice
- Antigen name:cAMP responsive element binding protein 1
- Cat. No.:ROCK100-401-195
- Reactivity:Human|Rat|Mouse
- Format:Antiserum
- Gene ID:1385
- Antigen synonyms:CREB
- Immunogen:CREB-1 (p43) peptide corresponding to a region near the N-terminus of the human Protein, conjugated to Keyhole Limpet Hemocyanin (KLH).
- Conjugation:Unconjugated
- Antigen symbol:CREB1
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- ELISA:Yes



