Order Entry
Poland
ContactUsLinkComponent
 
Anti-RFX5 Rabbit Polyclonal Antibody
  ROCK100-401-191
Anti-RFX5 Rabbit Polyclonal Antibody
  ROCK100-401-191

 

  • Antibody type:
    Primary
  • Antigen name:
    Regulatory factor X, 5 (influences HLA class II expression)
  • Antigen symbol:
    RFX5
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    near the C-terminus
  • Cross adsorption:
    none
  • Format:
    Antiserum
  • Form:
    Liquid (sterile filtered)
  • Gene ID:
    5993
  • Storage buffer:
    0,01% (w/v) Sodium Azide,
  • Storage temperature:
    Cool
  • Concentration:
    90 mg/ml by Refractometry
  • Shipping temperature:
    Ice
  • Immunogen:
    RFX5 (C-terminal specific) peptide corresponding to a region near the C-terminus of the human Protein conjugated to Keyhole Limpet Hemocyanin (KLH).
  • Purification:
    delipidation and defibrination
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

This product was assayed by immunoblot and found to be reactive against RFX5 (C-terminal specific) at a dilution of 1:250 followed by reaction with Peroxidase conjugated Affinity Purified anti-Rabbit IgG (H&L) (Goat).

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:3000;

Type: Primary
Antigen: RFX5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype:
Reactivity: Human