Order Entry
Poland
ContactUsLinkComponent
Anti-v-H-Ras Rat Monoclonal Antibody [clone: Y13-259]
Anti-v-H-Ras Rat Monoclonal Antibody [clone: Y13-259]
Catalog # OP01-100
Anti-v-H-Ras Rat Monoclonal Antibody [clone: Y13-259]
Catalog # OP01-100

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    V-H-RAS
  • Clonality:
    Monoclonal
  • Clone:
    Y13-259
  • Conjugation:
    Unconjugated
  • Host:
    Rat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Western blot:
    Yes
  • Neutralisation:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: v-H-Ras
Clonality: monoclonal
Clone: Y13-259
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: none
Reactivity: