Anti-His-Tag Mouse Monoclonal Antibody
Catalog # OB05-100
Supplier: Merck Millipore (Oncogene)
Specifications
- Antibody type:Primary
- Antigen symbol:His-Tag
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: His-Tag (His-Tag)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: