Order Entry
Poland
ContactUsLinkComponent
 
Anti-ANK3 Mouse Monoclonal Antibody
Catalog # NB20-100
Anti-ANK3 Mouse Monoclonal Antibody
Catalog # NB20-100

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ankyrin 3, Node Of Ranvier (ankyrin G)
  • Antigen symbol:
    ANK3
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: ANK3 (ankyrin 3, node of Ranvier (ankyrin G))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: