Anti-DNA-PK (Ab-2) (18-2) Mouse Monoclonal Antibody [clone: 42053]
NA57-100
- Antibody type:Primary
- Antigen symbol:DNA-PK
- Clonality:Monoclonal
- Clone:42053
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Neutralisation:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: DNA-PK (Ab-2) (18-2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human