Order Entry
Poland
ContactUsLinkComponent
Anti-CHEK1 Rabbit Polyclonal Antibody
Anti-CHEK1 Rabbit Polyclonal Antibody
Catalog # DR1025-50
Anti-CHEK1 Rabbit Polyclonal Antibody
Catalog # DR1025-50

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Checkpoint Kinase 1
  • Antigen symbol:
    CHEK1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µl
  • Pk:
    50 µl

Specifications

About this item

Type: Primary
Antigen: CHEK1 (checkpoint kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: