Anti-UPF1 Goat Polyclonal Antibody
Catalog # DR1015-100
Supplier: Merck Millipore (Oncogene)
Specifications
- Antibody type:Primary
- Antigen name:UPF1 regulator of nonsense transcripts homolog (yeast)
- Antigen symbol:UPF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: UPF1 (UPF1 regulator of nonsense transcripts homolog (yeast))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human