Order Entry
Poland
ContactUsLinkComponent
Anti-APCS Rabbit Polyclonal Antibody
Anti-APCS Rabbit Polyclonal Antibody
Catalog # 565191-1
Anti-APCS Rabbit Polyclonal Antibody
Catalog # 565191-1

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Amyloid P Component, Serum
  • Antigen symbol:
    Aminomethyltransferase
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    1 ml
  • Pk:
    1 mL

Specifications

About this item

Type: Primary
Antigen: APCS (amyloid P component, serum)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human