Anti-Nitrotyrosine Mouse Monoclonal Antibody [clone: CC22.8C7.3]
Catalog # 487923-50
Supplier: Merck Millipore (Oncogene)
Specifications
- Antibody type:Primary
- Antigen name:nitrotyrosine
- Clonality:Monoclonal
- Clone:CC22.8C7.3
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Multiple
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: Nitrotyrosine (nitrotyrosine)
Clonality: monoclonal
Clone: CC22.8C7.3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Pan