Anti-Heme Oxygenase-1 (1-30) Rabbit Polyclonal Antibody
374090-100
- Antibody type:Primary
- Antigen name:Heme Oxygenase-1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µG
Type: Primary
Antigen: Heme Oxygenase-1 (1-30)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: