Anti-Dopamine D 2S Receptor Rabbit Polyclonal Antibody
324396-100
- Antibody type:Primary
- Antigen name:Dopamine D2S receptor
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: Dopamine D 2S Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: