Anti-alpha-Actin (1A4) Mouse Monoclonal Antibody
Catalog # 113200-500
Supplier: Merck Millipore (Oncogene)
Specifications
- Antibody type:Primary
- Antigen name:alpha Actin
- Antigen symbol:1A4
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:500 µl
- Pk:500 µl
Specifications
About this item
Type: Primary
Antigen: alpha-Actin (1A4)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: