Order Entry
Poland
ContactUsLinkComponent
 
Anti-Growth hormone Receptor Rabbit Polyclonal Antibody
  BSBTPA1726
undefined
Anti-Growth hormone Receptor Rabbit Polyclonal Antibody
  BSBTPA1726

 

  • Antibody type:
    Primary
  • Antigen name:
    Growth hormone receptor
  • Antigen symbol:
    HGHR
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Amino acid number:
    231 to 247
  • Sequence:
    DKEHEVRVRSRQRSFEK
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence in the middle region of mouse Growth hormone receptor (231-247aa DKEHEVRVRSRQRSFEK), identical to the related rat sequence.
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

 

 

Rabbit IgG polyclonal antibody for Growth hormone receptor(GHR) detection. Tested with WB in Mouse; Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Growth hormone Receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat