Specifications
- Antibody type:Primary
- Antigen name:Mucin Gastric
- Antigen symbol:Mucin Gastric
- Clonality:Monoclonal
- Clone:MG-31
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Rabbit,Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilized
- Gene ID:Human(4586), Mouse(17833), Rat(682837)
- Antigen synonyms:Mucin-5AC
- UniProtKB:Human(P98088)
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:Mucin from human ovarian cyst fluid.
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Mouse IgG monoclonal antibody for Mucin Gastric, mucin 5AC, oligomeric mucus/gel-forming (MUC5AC) detection. Tested with WB, IHC-P, ICC in Human. No cross reactivity with other proteins.
Mucin 5AC (Muc5AC) is a protein that in humans is encoded by the MUC5AC gene. Using real-time RT-PCR, Moehle et al. (2006) found that MUC5AC was highly expressed in adult trachea and stomach. Expression was lower in adult lung and weak in fetal lung, and no expression was detected in other tissues. This gene has been linked to mucus hypersecretion in the pulmonary tracts and associated to chronic obstructive pulmonary disease. Moreover, Muc5ac is identified as a direct and critical mediator of resistance during intestinal nematode infection.
1.2% sodium acetate or neutral PBS. If 1ml of PBS is used, the antibody concentration will be 100ug/ml.
Mouse ascites fluid, 1.2% sodium acetate, 2mg BSA, with 0.01mg NaN3 as preservative.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: Mucin Gastric
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Rabbit