Anti-IGF1 Receptor Rabbit Polyclonal Antibody
Catalog # BSBTPA1746
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Insulin-Like Growth Factor 1 Receptor
- Antigen symbol:IGF1R
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:MGC18216|JTK13|CD221|IGFIR|MGC142170|IGF1R|MGC142172
- Amino acid number:1353 - 1367
- Sequence:RKNERALPLPQSSTC
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human IGF1 Receptor (1353-1367aa RKNERALPLPQSSTC), different from the related rat and mouse sequences by one amino acid.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Insulin-like growth factor 1 receptor (IGF1R) detection. Tested with WB, IHC-P in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: IGF1 Receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat