Order Entry
Poland
ContactUsLinkComponent
Anti-XRCC3 Rabbit Polyclonal Antibody
Anti-XRCC3 Rabbit Polyclonal Antibody
Catalog # BSBTPA1697
Supplier:  Boster Bio
undefined
Anti-XRCC3 Rabbit Polyclonal Antibody
Catalog # BSBTPA1697
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    X-ray Repair Complementing Defective Repair in Chinese Hamster cells 3
  • Antigen symbol:
    XRCC3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    X ray repair complementing defective repair in Chinese hamster cells 3|CMM6|X ray repair cross complementing protein 3|DNA repair protein XRCC3
  • Amino acid number:
    290 to 305
  • UniProtKB:
    Q9CXE6
  • Sequence:
    NQLLMRLMVDRTHEDD
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of mouse XRCC3 (290-305aa NQLLMRLMVDRTHEDD), identical to the related rat sequence.
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for DNA repair protein XRCC3(XRCC3) detection. Tested with WB, IHC-P in Mouse;Rat.

XRCC3(X-RAY REPAIR, COMPLEMENTING DEFECTIVE, IN CHINESE HAMSTER, 3) is a DNA repair protein that in humans, is encoded by the XRCC3 gene. The XRCC3 is a member of the RecA/Rad51-related protein family that participates in homologous recombination to maintain chromosome stability and repair DNA damage. XRCC3 interacts directly with RAD51 and may cooperate with RAD51 during recombinational repair. XRCC3 function is not limited to HR initiation, but extends to later stages in formation and resolution of HR intermediates, possibly by stabilizing heteroduplex DNA. The gene functionally complements Chinese hamster irs1SF, a repair-deficient mutant that exhibits hypersensitivity to a number of different DNA-damaging agents and is chromosomally unstable. A rare microsatellite polymorphism in this gene is associated with cancer in patients of varying radiosensitivity.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.

Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: XRCC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat